Order Entry
Italy
Orders LinkContactUsLinkComponent
Dip slides
Dip slides
V.R.B.G. agar / Rose Bengal, 20
Catalog # LIOF525202
Dip slides
Catalog # LIOF525202
Supplier:  
CAS Number:  
Dip slides
Dip slides
V.R.B.G. agar / Rose Bengal, 20
CAS Number:  
Discontinued
Product Details & Documents
Flexible contact slide for microbiology screening of surfaces.
  • ISO 18593
Documents
Specifications
Specifications
  • Pk:
    20
  • Description:
    V.R.B.G. agar / Rose Bengal