Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Destain solutions

Destain I (45% Methanol, 9% Glacial acetic acid), 1 L
Catalog # 786-526
Supplier:   G-Biosciences
Destain solutions
Catalog # 786-526
Supplier:  G-Biosciences
CAS Number:  
Destain solutions
Destain solutions

Destain I (45% Methanol, 9% Glacial acetic acid), 1 L

CAS Number:  
Price
84,70 €
Log in for availability
1 * 1 L
Product Details & Documents

G-Biosciences' Destain I contains approximately 45% methanol and 9% glacial acetic acid. G-Biosciences' Destain II contains approximately 5% methanol and 7% acetic acid.

Specifications

Specifications

  • Pk:
    1 L
  • Description:
    Destain I (45% Methanol, 9% Glacial acetic acid)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.