Order Entry
Italy
Orders LinkContactUsLinkComponent
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Automatic pressure controller for 230 V units, incl. mounting bracket, hardware, 3/16" ID polyvinyl tubing, 3/32" ID polyvinyl tubing, and power cord, for automatic control of the main chamber pressure, main chamber purging and filling cycles, and transfer chamber evacuation and filling cycles
Catalog # 135-1088
Supplier:   LABCONCO
Accessories for glove boxes, Precise™
Catalog # 135-1088
Supplier:  LABCONCO
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Automatic pressure controller for 230 V units, incl. mounting bracket, hardware, 3/16" ID polyvinyl tubing, 3/32" ID polyvinyl tubing, and power cord, for automatic control of the main chamber pressure, main chamber purging and filling cycles, and transfer chamber evacuation and filling cycles
Price
9 490,00 €
Log in for availability
1 * 1 pezzi
Product Details & Documents
Specifications
  • Description:
    Automatic pressure controller for 230 V units, incl. mounting bracket, hardware, 3/16" ID polyvinyl tubing, 3/32" ID polyvinyl tubing, and power cord, for automatic control of the main chamber pressure, main chamber purging and filling cycles, and transfer chamber evacuation and filling cycles
  • Seq:
    01