Order Entry
Italy
Orders LinkContactUsLinkComponent
Accessories for glove boxes, Precise™
Viewing window, acrylic (749×396×12,7 mm) with dry powder epoxy-coated steel frame, replaces standard laminated glass window
Catalog # LBCP5236100
Supplier:   LABCONCO
Accessories for glove boxes, Precise™
Catalog # LBCP5236100
Supplier:  LABCONCO
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Viewing window, acrylic (749×396×12,7 mm) with dry powder epoxy-coated steel frame, replaces standard laminated glass window
Price
1 010,00 €
Log in for availability
1 * 1 pezzi
Product Details & Documents
Specifications
  • Description:
    Viewing window, acrylic (749×396×12,7 mm) with dry powder epoxy-coated steel frame, replaces standard laminated glass window
  • Seq:
    36