Order Entry
Italy
ContactUsLinkComponent
Stirring-heating plate with 6 stirring positions, universally applicable for miscellaneous vessel sizes and shapes, stirring point d istance 130 mm, suitable for 6 x 1,000 ml Erlenmeyer flasks. 1 * 1 items
Catalog # 442-0761
Supplier:  2MAG
Stirring-heating plate with 6 stirring positions, universally applicable for miscellaneous vessel sizes and shapes, stirring point d istance 130 mm, suitable for 6 x 1,000 ml Erlenmeyer flasks. 1 * 1 items
Catalog # 442-0761
Supplier:  2MAG
Frequently Bought Together