Order Entry
Italy
ContactUsLinkComponent
Magnetic stirrer, miniaturized, wear-free with 1 stirring position, for up to 25 ml stirring volume.1 stirring position, 100% mainte nance-free and wear-free by inductive 2mag- 1 * 1 items
Catalog # TMAG40400_CH
Supplier:  2MAG
Magnetic stirrer, miniaturized, wear-free with 1 stirring position, for up to 25 ml stirring volume.1 stirring position, 100% mainte nance-free and wear-free by inductive 2mag- 1 * 1 items
Catalog # TMAG40400_CH
Supplier:  2MAG
Frequently Bought Together