Order Entry
Italy
Orders LinkContactUsLinkComponent
Visual colour kits, zinc, MQuant®, Supelco®

Visual colour kits, zinc, MQuant®, Supelco®

Zinc test, 0,1-0,2-0,3-0,4-0,5-0,7-1-2-5 mg/l Zn, Thiocyanate/ brilliant green, MCD, 120 Tests
Codice catalogo 1.14780.0001
Nome del fornitore:   Merck
Visual colour kits, zinc, MQuant®, Supelco®
Codice catalogo 1.14780.0001
Nome del fornitore:  Merck
Numero CAS:  
Visual colour kits, zinc, MQuant®, Supelco®
Visual colour kits, zinc, MQuant®, Supelco®

Zinc test, 0,1-0,2-0,3-0,4-0,5-0,7-1-2-5 mg/l Zn, Thiocyanate/ brilliant green, MCD, 120 Tests

Numero CAS:  
Price
547,00 €
Log in for availability
1 * 120 Tests
Product Details & Documents

Quick tests, visual colour comparison.

  • Results in approximately five minutes
  • Easy to use
  • Application-oriented measuring ranges
  • Portable
  • A range of economical refill packs (RP) are available

MQuant® with colour card comparator

By using cuvettes with a long light path, the MQuant® (MCC) test kits are especially suited for measuring low ion concentrations (some as low as 0,005 mg/L). These test kits are especially appropriate for analysing drinking water, spring water, purified and mineral water, as well as cooling and boiler water.

MQuant® with colour disc comparator MCD

Using the comparator disc, these test kits are especially suitable for the analysis of turbid and coloured waters. The comparator disc is a practical and robust colour comparison device that provides reliable measurements, even in humid environments. Ideal for samples of medium to high concentration in applications ranging from testing bottled water to wastewater and other industrial water testing.

Delivery information: Each test kit contains all the necessary reagents, reaction cuvettes, colour comparison chart or comparator wheel and extensive instructions for use.

Specifications

Specifications

  • Chemical method:
    Thiocyanate/ brilliant green
  • Pk:
    120 Tests
  • Description:
    Zinc test
  • Type:
    MCD
  • Range:
    0,1-0,2-0,3-0,4-0,5-0,7-1-2-5 mg/l Zn

Specifications