Order Entry
Italy
Orders LinkContactUsLinkComponent
Selfpacker® Preparative HPLC Columns
Selfpacker® Preparative HPLC Columns
Complete column tube with temperature control jacket and top flange chain bolted without hydraulics, — mm, — mm
  556-0023
Selfpacker® Preparative HPLC Columns
  556-0023
Selfpacker® Preparative HPLC Columns
Selfpacker® Preparative HPLC Columns
Complete column tube with temperature control jacket and top flange chain bolted without hydraulics, — mm, — mm
1 * 1 pezzi
This column-packing technology is based on a combination of vacuum suction with subsequent column bed compression that stabilises the packing. A common water-jet pump is used to generate the vacuum. The sorbent slurry is sucked through a packing reservoir into the column by applying low pressure at the bottom end of the column.
  • Allows precise adjustment of the desired packing volume or sorbent bed height
  • Facilitates the determination of the column geometry
  • Makes direct scale-up from analytical to preparative scale easy
Packing stand NW25
Min./max. packing height: 185/220 mm
Max. working pressure: 200 bar
Packing stand NW50
Min./max. packing height: 185/220 mm
Max. working pressure: 100 bar
Packing stand NW100
Min./max. packing height: 295/350 mm
Max. working pressure: 100 bar
Max. working pressure: 200 bar
  • Ø int.:
    — mm
  • Description:
    Complete column tube with temperature control jacket and top flange chain bolted without hydraulics
  • Length:
    — mm
  • Seq:
    34