Copper etchant
3,8 L
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Applications: Iron (III) chloride solution for immersion etching copper, copper alloys and Kovar in printed circuit board applications
Physical Form: Liquid Density: 1.19 Appearance: Dark yellow to brown Odour: Acidic UN No.: UN3264 MDL No.: MFCD00161912
Applications: Iron (III) chloride solution for immersion etching copper, copper alloys and Kovar in printed circuit board applications
Synonyms: Copper etching solution
- MDL Number:MFCD00161912
- Density:1.19
- Storage Temperature:Ambient
- UN:3264
- ADR:8,III
Documents
Specifications
Specifications
- Pk:3,8 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



