Order Entry
Italy
Orders LinkContactUsLinkComponent
CF® Dye hydrazide
CF® Dye hydrazide
CF®647 hydrazide, 650/665 nm, 1 mg
Catalog # 92136.
Supplier:   Biotium
CF® Dye hydrazide
Catalog # 92136.
Supplier:  Biotium
CAS Number:  
CF® Dye hydrazide
CF® Dye hydrazide
CF®647 hydrazide, 650/665 nm, 1 mg
CAS Number:  
Price
209,00 €
Log in for availability
1 * 1 mg
Product Details & Documents
Fluorescent CF® Dye hydrazides are bright, extremely water-soluble, nontoxic molecules, useful as aldehyde-fixable cell tracers. They can also be used to label aldehyde or ketone groups of polysaccharides, carbohydrates or glycoproteins.
  • Fluorescently labeled low molecular weight, polar, non-toxic molecules; useful as aldehyde-fixable cell tracers
  • Membrane impermeant, can be easily loaded into cells by microinjection
  • Can be used to fluorescently label aldehyde or ketone groups of polysaccharides, carbohyrdates, or glycoproteins
  • Bright, photostable and water-soluble CF® Dyes are excellent options for fluorescent imaging
Bright, extremely water-soluble and non-toxic CF® Dye hydrazides are excellent fluorescent tracers for neurons or gap junction studies. The cell impermeant tracer dyes can be introduced into cells by microinjection.
CF® Dye hydrazides can also be used to fluorescently label aldehyde or ketone groups of carbohydrates or glycoproteins molecules after peroxidation with periodate.
For carbohydrates labeling applications, we recommend using CF® Dye aminooxy forms, which are more reactive toward these groups than hydrazide forms. For fluorescence two-dimensional differential gel electrophoresis (2D-DIGE), we recommend CFDI hydrazides.
Specifications
Specifications
  • Pk:
    1 mg
  • Description:
    CF®647 hydrazide
  • Ex/Em:
    650/665 nm