Order Entry
Italy
Orders LinkContactUsLinkComponent
VWR® Basic Cleanroom Lab Coats, SPP, Disposable

VWR® Basic Cleanroom Lab Coats, SPP, Disposable

White, XL, 110 cm
Catalog # 113-1121
Supplier:   VWR Collection
VWR® Basic Cleanroom Lab Coats, SPP, Disposable
Catalog # 113-1121
Supplier:  VWR Collection
VWR® Basic Cleanroom Lab Coats, SPP, Disposable
VWR® Basic Cleanroom Lab Coats, SPP, Disposable

White, XL, 110 cm

Price
71,60 €
Log in for availability
1 * 25 pezzi
Product Details & Documents

VWR® Basic SPP lab coats are manufactured from a strong, lightweight and breathable spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates in non hazardous environments.

  • Tapered collar, inset sleeves, snap closure
  • Elasticated or knitted cuffs, no pockets or three pockets
  • Lightweight and breathable first line barrier protection

This garment is ideal for less critical areas or pre-gowning entry rooms. It is tested and manufactured in an ISO certified facility and lot controlled and validated through independent laboratory testing.

Packaging: 25 or 30 coats per carton.

Specifications
  • Clothing Size:
    XL
  • Colour:
    White
  • Length:
    110 cm
  • Seq:
    06
  • Cat. No.:
    113-1121
  • Garment application:
    Cleanroom|Laboratory|Industrial
  • Gender:
    Unisex
  • Pockets:
    Without pockets
  • Closure type:
    Press studs / snaps
  • Weight:
    45 g/m²
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.