Order Entry
Italy
Orders LinkContactUsLinkComponent
Dish-shaped Dewar flasks, type SCH-CAL
Dish-shaped Dewar flasks, type SCH-CAL
Dewar flask SCH 18 CAL
Catalog # KGWA1673
Supplier:   KGW
Dish-shaped Dewar flasks, type SCH-CAL
Catalog # KGWA1673
Supplier:  KGW
Dish-shaped Dewar flasks, type SCH-CAL
Dish-shaped Dewar flasks, type SCH-CAL
Dewar flask SCH 18 CAL
Price
58,30 €
Log in for availability
1 * 1 pezzi
Product Details & Documents
Silvered borosilicate glass, structured aluminium protective covering. For tempering of round bottom flasks, cooling of small samples. They are vacuum insulated and silvered.
  • Suitable as hot or cold bath
  • Can be used on a magnetic stirrer
  • The cover is anti-magnetic and stainless
Temperature range from −200 to +200 °C.
CAL flasks have an external stucco aluminium protective casing, alternatively the glass inserts (without casing) are also available.
Specifications
  • Seq:
    11
  • For:
    Dewar flask SCH 18 CAL