Order Entry
Italy
Orders LinkContactUsLinkComponent
Globe stirrer couplings, BOLA

Globe stirrer couplings, BOLA

Stirrer coupling for shafts with Ø 6,5 or 10 mm opening, with Ø top 10 mm, , total length 190 mm
Codice catalogo BOHLC398-08
Nome del fornitore:   Bohlender
Globe stirrer couplings, BOLA
Codice catalogo BOHLC398-08
Nome del fornitore:  Bohlender
Globe stirrer couplings, BOLA
Globe stirrer couplings, BOLA

Stirrer coupling for shafts with Ø 6,5 or 10 mm opening, with Ø top 10 mm, , total length 190 mm

Price
154,00 €
Log in for availability
1 * 1 pezzi
Product Details & Documents

These POM couplings are ideal for balancing misalignment of axes between stirrer and stirrer shaft. Suitable for glass, metal or Bola stirrer shafts. Both models are suitable for rotational speeds of up to 1200 rpm and can be used for both right and left-handed rotation.

  • Excellent chemical resistance, temperature resistance from –20 to +110 °C
  • Assembled by means of screw joints with clamp rings
  • Pivot (length 90 mm) can be shortened by the user
  • Maximum misalignment of 10 mm

Powerful transmission of up to 300 Ncm.

Powerful transmission of up to 300 Ncm.

Specifications
  • Description:
    Stirrer coupling for shafts with Ø 6,5 or 10 mm opening, with Ø top 10 mm, , total length 190 mm
  • Seq:
    04
Accessories

Product Information

  • Codice catalogo
    Description
    Disponibilità
    Price
  • BOHLC911-02
    Replacement-reducing sleeve, PTFE glass fibre
    Log in for availability
    1 * 3 pezzi - 15,80 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.