Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Solid phase extraction cartridges, HyperSep™ SCX

Cartridge, SCX, 40 - 60 µm, 6 ml, 1000 mg
Catalog # HYPE60108-433
Solid phase extraction cartridges, HyperSep™ SCX
Catalog # HYPE60108-433
Solid phase extraction cartridges, HyperSep™ SCX
Solid phase extraction cartridges, HyperSep™ SCX

Cartridge, SCX, 40 - 60 µm, 6 ml, 1000 mg

Price
241,00 €
Log in for availability
1 * 30 pezzi
Product Details & Documents

A strong cation exchange sorbent for extraction of charged basic compounds.

  • Benzene ring provides potential for nonpolar interactions
  • Low pKa value sorbent
Specifications
  • Column Format:
    Cartridge
  • Particle Size:
    40 - 60 µm
  • Capacity:
    6 ml
  • Seq:
    06
  • Bed weight:
    1000 mg
  • Phase:
    SCX