Order Entry
Italy
Orders LinkContactUsLinkComponent
Starter-Kit, SL 1011

Starter-Kit, SL 1011

SI 2150, Interface IF 150, software Winlog.med
Catalog # EBRO1340-6193
Supplier:   EBRO
Starter-Kit, SL 1011
Catalog # EBRO1340-6193
Supplier:  EBRO
Starter-Kit, SL 1011
Starter-Kit, SL 1011

SI 2150, Interface IF 150, software Winlog.med

Price
2 490,00 €
Log in for availability
1 * 1 pezzi
Product Details & Documents

The SL1011 set by ebro, featuring the EBI 12-T101 data logger, PHX800 pH tester, and TDS3 conductivity meter, is designed for validating and monitoring bedpan washers in healthcare settings, ensuring hygiene standards.

  • Comprehensive validation set for bedpan washers
  • Includes EBI 12-T101 temperature data logger, PHX800 pH tester, and TDS3 conductivity meter
  • High-precision monitoring for temperature, pH, and conductivity
  • Ideal for healthcare hygiene checks
  • Compact and durable instruments with high IP protection
  • Traceable factory calibration certificate at –20 °C, 0 °C and +60 °C

The SL1011 set by ebro is a complete solution for monitoring bedpan washers and washers and disinfectors crucial in healthcare environments to ensure compliance with hygiene standards. This set includes the EBI 12-T101 temperature data logger, PHX800 pH tester, and TDS3 conductivity tester, covering critical parameters: temperature, pH, and conductivity. The EBI 12-T101 accurately logs temperature data, while the PHX800 and TDS3 provide reliable pH and conductivity measurements, ensuring comprehensive quality checks. The instruments are compact, robust, and user-friendly, designed to handle frequent use and withstand challenging conditions.

Dimensions: Ø 48×24 mm H

Specifications
  • Description:
    SI 2150, Interface IF 150, software Winlog.med
  • Seq:
    03
Complementary Products

Product Information

  • Catalog #
    Accuracy
    Description
    Range
    Resolution
    Availability
    Price
  • EBRO1340-6600
    ±0,5 °C (–90…–40 °C); ±0,2 °C (–40…0 °C); ±0,1 °C (0…120 °C); ±0,05 °C (120…140 °C); ±0,1 °C (140…150 °C)
    EBI 12-T100, high temperature data logger, pharmaceutical, medical and food applications; calibration points: –20, 0, 60 and 134 °C
    T: –90…+150 °C
    0,01 °C
    Log in for availability
    1 * 1 pezzi - 1 920,00 €

  • EBRO1340-6637
    ±0,1 °C (0…140 °C)
    ±0,2 °C (–40…0 °C)
    EBI 12-T237, high temperature data logger
    T: –40…+140 °C
    0,01 °C
    Log in for availability
    1 * 1 pezzi - 1 890,00 €

  • EBRO1340-6658
    ±0,1 (0…140 °C)
    ±20 mbar
    EBI 12-TP237, temperature and pressure data logger, calibration points 0 °C, +60 °C and +134 °C; 100 mbar and 3100 mbar (bei +25 °C); 3100 mbar (at +134 °C)
    T: 0…140 °C
    P: 1…4000 mbar
    0,01 °C
    1 mbar
    Log in for availability
    1 * 1 pezzi - 2 330,00 €

Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Battery replacement set EBI 12; 3× battery, 6× O-ring
    Battery replacement set EBI 12; 3× battery, 6× O-ring
    620-1693
    Log in for availability
    1 * 1 pezzi - 185,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Battery replacement set EBI 12; 3× battery, 6× O-ring, 1× unscrewing aid EBI 12
    Battery replacement set EBI 12; 3× battery, 6× O-ring, 1× unscrewing aid EBI 12
    EBRO1100-0130
    Log in for availability
    1 * 1 pezzi - 264,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.