Order Entry
Italy
Orders LinkContactUsLinkComponent
Consumables for infrared spectroscopy
Consumables for infrared spectroscopy
Sodium chloride (NaCl) circular demountable cell windows, one pair, thickness: 4 mm
Catalog # PERCL1271102
Supplier:   PerkinElmer
Consumables for infrared spectroscopy
Catalog # PERCL1271102
Supplier:  PerkinElmer
Consumables for infrared spectroscopy
Sodium chloride (NaCl) circular demountable cell windows, one pair, thickness: 4 mm
Price
211,00 €
Log in for availability
1 * 2 pezzi
Product Details & Documents
Specifications
  • Description:
    Sodium chloride (NaCl) circular demountable cell windows, one pair, thickness: 4 mm
  • Seq:
    19