Order Entry
Italy
Orders LinkContactUsLinkComponent
Single channel pipettes, mechanical, fixed / variable volume, labopette®
50 µl, ±1,0 %, <0,4 %, Yellow, 1 - 200 µl
Catalog # HIRS9475505
Supplier:   HIRSCHMANN
Single channel pipettes, mechanical, fixed / variable volume, labopette®
Catalog # HIRS9475505
Supplier:  HIRSCHMANN
Single channel pipettes, mechanical, fixed / variable volume, labopette®
Single channel pipettes, mechanical, fixed / variable volume, labopette®
50 µl, ±1,0 %, <0,4 %, Yellow, 1 - 200 µl
Price
251,00 €
Log in for availability
1 * 1 pezzi
Product Details & Documents
The reliable labopette® pipette features superior ergonomic characteristics, right down to the smallest detail. The entire working process is optimised to ensure non fatiguing work that places a minimum of stress on the joints.
  • Volume configuration with digital locking feature
  • With tip ejection and calibration option
  • Tip seal and safety filter to prevent contamination
  • Trigger grip ensures a firm but relaxed hold during pipetting
  • Ergonomic design of the handle offers an equally firm, comfortable hold for large and small hands, right and left-handed users and hands with or without gloves
  • Effective thermal insulation of the unit interior prevents hand heat penetrating into the interior of the unit
  • Completely autoclavable without dismantling
 : Supplied with calibration tool, pipette holder, individual serial number and calibration certificate.
Specifications
  • Volume:
    50 µl
  • Seq:
    13
  • Accuracy:
    ±1,0 %
  • Imprecision:
    <0,4 %
  • Recommended tips:
    Yellow, 1 - 200 µl
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Safe-Cone filters, for labopette® variable 1000 - 5000 µl; fix 2000/5000 µl
    Safe-Cone filters, for labopette® variable 1000 - 5000 µl; fix 2000/5000 µl
    HIRS9470010
    Log in for availability
    1 * 50 pezzi - 58,90 €
Product Information
  • Catalog #
    Availability
    Price
  • Safe-Cone filters, for labopette® variable 2 - 20/5 - 50 µl; fix 20/25/50 µl
    Safe-Cone filters, for labopette® variable 2 - 20/5 - 50 µl; fix 20/25/50 µl
    HIRS9470013
    Log in for availability
    1 * 50 pezzi - 58,90 €
Product Information
  • Catalog #
    Availability
    Price
  • Safe-Cone filters, for labopette® elctronic 10 - 500/50 - 1000/500 - 5000/50 - 1200 µl; variable 100 - 1000 µl; fix 250/500/1000 µl
    Safe-Cone filters, for labopette® elctronic 10 - 500/50 - 1000/500 - 5000/50 - 1200 µl; variable 100 - 1000 µl; fix 250/500/1000 µl
    HIRS9470011
    Log in for availability
    1 * 50 pezzi - 58,90 €
Product Information
  • Catalog #
    Availability
    Price
  • Safe-Cone filters, for labopette® variable 10 - 100/20 - 200 µl; fix 100/200 µl
    Safe-Cone filters, for labopette® variable 10 - 100/20 - 200 µl; fix 100/200 µl
    HIRS9470012
    Log in for availability
    1 * 50 pezzi - 58,90 €
Product Information
  • Catalog #
    Availability
    Price
  • DKD calibration certificate for labopette® fix 0,5 μl - 5 ml
    DKD calibration certificate for labopette® fix 0,5 μl - 5 ml
    HIRS9980116
    Log in for availability
    1 * 1 pezzi - 91,00 €
Product Information
  • Catalog #
    Availability
    Price
  • DKD calibration certificate for labopette® 12-channel, 5 - 250 μl
    DKD calibration certificate for labopette® 12-channel, 5 - 250 μl
    HIRS9980118
    Log in for availability
    1 * 1 pezzi - 395,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Stand for six mechanical pipettes
    Stand for six mechanical pipettes
    HIRS9470006
    Log in for availability
    1 * 1 pezzi - 86,60 €
Product Information
  • Catalog #
    Availability
    Price
  • Carousel stand for six mechanical pipettes
    Carousel stand for six mechanical pipettes
    HIRS9470004
    Log in for availability
    1 * 1 pezzi - 175,00 €
Product Information
  • Catalog #
    Volume
    Tip colour
    For use with
    Availability
    Price
  • HIRS9490101
    0,2 - 10 µl
    Transparent
    Hirschmann Laborgeräte, Gilson® pipettes
    Log in for availability
    1 * 1.000 pezzi - 34,00 €
Product Information
  • Catalog #
    Volume
    Tip colour
    For use with
    Availability
    Price
  • HIRS9490102
    1 - 200 µl
    Yellow
    Hirschmann Laborgeräte, Eppendorf®, Brand®, Gilson® pipettes
    Log in for availability
    1 * 1.000 pezzi - 22,70 €
Product Information
  • Catalog #
    Volume
    Tip colour
    For use with
    Availability
    Price
  • HIRS9490103
    100 - 1000 µl
    Blue
    Hirschmann Laborgeräte, Eppendorf®, Brand®, Gilson® pipettes
    Log in for availability
    1 * 1.000 pezzi - 25,30 €
Product Information
  • Catalog #
    Volume
    Tip colour
    For use with
    Availability
    Price
  • HIRS9490106
    1 - 250 µl
    Transparent
    labopette® multichannel pipettes
    Log in for availability
    1 * 1.000 pezzi - 36,80 €
Product Information
  • Catalog #
    Volume
    Tip colour
    For use with
    Availability
    Price
  • HIRS9490124
    100 - 5000 µl
    Transparent
    Hirschmann Laborgeräte pipettes
    Log in for availability
    1 * 300 pezzi - 54,00 €
Product Information
  • Catalog #
    Volume
    Tip colour
    For use with
    Availability
    Price
  • HIRS9490201
    0,2 - 10 µl
    Transparent
    Hirschmann Laborgeräte, Gilson® pipettes
    Log in for availability
    1 * 960 pezzi - 81,00 €
Product Information
  • Catalog #
    Volume
    Tip colour
    For use with
    Availability
    Price
  • HIRS9490202
    1 - 200 µl
    Yellow
    Hirschmann Laborgeräte, Eppendorf®, Brand®, Gilson® pipettes
    Log in for availability
    1 * 960 pezzi - 61,10 €
Product Information
  • Catalog #
    Volume
    Tip colour
    For use with
    Availability
    Price
  • HIRS9490205
    50 - 1200 µl
    Transparent
    Hirschmann Laborgeräte pipettes
    Log in for availability
    1 * 960 pezzi - 151,00 €
Product Information
  • Catalog #
    Volume
    Tip colour
    For use with
    Availability
    Price
  • HIRS9490206
    1 - 250 µl
    Transparent
    labopette® multichannel pipettes
    Log in for availability
    1 * 960 pezzi - 60,70 €
Product Information
  • Catalog #
    Volume
    Tip colour
    For use with
    Availability
    Price
  • HIRS9490104
    1 - 10000 µl
    Transparent
    Hirschmann Laborgeräte pipettes
    Log in for availability
    1 * 100 pezzi - 54,00 €
Product Information
  • Catalog #
    Tip style
    For use with
    Availability
    Price
  • HIRS9490109
    Multibox, autoclavable
    refill trays of pipette tips up to 300 µl
    Log in for availability
    1 * 1 pezzi - 29,00 €
Product Information
  • Catalog #
    Tip style
    For use with
    Availability
    Price
  • HIRS9490110
    Multibox, autoclavable
    refill trays of pipette tips up to 1000 µl
    Log in for availability
    1 * 1 pezzi - 30,60 €
Product Information
  • Catalog #
    Availability
    Price
  • Safe-Cone filters, for labopette® fix 10/20/25/250 µl; multi-channel 5,0 - 50/50 - 300 µl; electronic multi-channel 25 - 250/5 - 100 µl
    Safe-Cone filters, for labopette® fix 10/20/25/250 µl; multi-channel 5,0 - 50/50 - 300 µl; electronic multi-channel 25 - 250/5 - 100 µl
    HIRS9470014
    Log in for availability
    1 * 50 pezzi - 58,90 €
Product Information
  • Catalog #
    Availability
    Price
  • DKD calibration certificate for labopette® variable volumes 0,5 μl - 5 ml / labopettor
    DKD calibration certificate for labopette® variable volumes 0,5 μl - 5 ml / labopettor
    HIRS9980115
    Log in for availability
    1 * 1 pezzi - 147,00 €
Product Information
  • Catalog #
    Availability
    Price
  • DKD calibration certificate for labopette® 8-channel, 0,5 - 250 μl
    DKD calibration certificate for labopette® 8-channel, 0,5 - 250 μl
    HIRS9980117
    Log in for availability
    1 * 1 pezzi - 352,00 €