Order Entry
Italy
ContactUsLinkComponent
Adapter glass filled ptfe rodaviss joint npt 1 1/2 taper 34 ht55x48di
  COWIPC712.3438.GF
 :  COWIE
Adapter glass filled ptfe rodaviss joint npt 1 1/2 taper 34 ht55x48di
  COWIPC712.3438.GF
 :  COWIE