Order Entry
Italy
Orders LinkContactUsLinkComponent
L(+)-Lysine monohydrate

L(+)-Lysine monohydrate

Glass bottle for solids, 100 g
Catalog # A1342.0100
Supplier:   PanReac AppliChem
L(+)-Lysine monohydrate
Catalog # A1342.0100
Supplier:  PanReac AppliChem
CAS Number:  39665-12-8
L(+)-Lysine monohydrate

Glass bottle for solids, 100 g

CAS Number:  39665-12-8
Price
192,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  L-(+)-Lysine, Hydrate, (S)-(+)-2,6-Diaminohexanoic acid monohydrate, (S)-(+)-Lysine monohydrate, L-Lysine hydrate, (S)-(+)-2,6-Diaminocaproic acid monohydrate, H-Lys-OH monohydrate, K monohydrate, H-Lys-OH·H₂O, Lys monohydrate
  • CAS Number:39665-12-8
  • Formula:H₂N(CH₂)₄CH(NH₂)CO₂H·H₂O
  • MDL Number:MFCD00151035
  • Molecular Weight:164.2 g/mol
  • Melting Pt:225 °C
  • Storage Temperature:Refrigerator
  • EINECS:200-294-2

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Cat. No.:
    A1342.0100

Specifications

Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..