Order Entry
Italy
ContactUsLinkComponent
MULTIMETER DIGITAL MULTI 3430 SET G : Professional digital multi meter for mobile measurement, triple channel input, colour graphic display including datalogger and USB interfaces. Multi case set with IDS sensors: digital pH sensor SenTix 940-3, Tetr 1 * 1 items
Catalog # 665-0234
Nome del fornitore:  WTW
MULTIMETER DIGITAL MULTI 3430 SET G : Professional digital multi meter for mobile measurement, triple channel input, colour graphic display including datalogger and USB interfaces. Multi case set with IDS sensors: digital pH sensor SenTix 940-3, Tetr 1 * 1 items
Catalog # 665-0234
Nome del fornitore:  WTW
Frequently Bought Together