Order Entry
Italy
Orders LinkContactUsLinkComponent
Funnels with GL thread or ground joint

Funnels with GL thread or ground joint

20 mm
Codice catalogo BOHLD739-22
Nome del fornitore:   Bohlender
Funnels with GL thread or ground joint
Codice catalogo BOHLD739-22
Nome del fornitore:  Bohlender
Funnels with GL thread or ground joint
Funnels with GL thread or ground joint

20 mm

Price
200,00 €
Log in for availability
1 * 1 pezzi
Product Details & Documents

Powder funnels, made of borosilicate glass with outlet pipe of PTFE, connection with GL screw caps of PPS or ground joint.

  • The glass funnel can be fixed in any position
  • Used for reaction vessels with GL threaded connections or grounding sleeves
  • Temperature resistance −20 to +200 °C
  • Universal chemical resistance
Specifications
  • Stem O-Ø:
    20 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.