Accessories for heating circulators
Stainless steel PT100 sensor with 3 m cable, 900×6 mm, CF31, CF41, F25-HE, F25-HL, F25-ME
Catalog # JULA8981016
Supplier: Julabo GmbH
Jump to:
Product Details & Documents
Specifications
- Description:Stainless steel PT100 sensor with 3 m cable, 900×6 mm
- For:CF31, CF41, F25-HE, F25-HL, F25-ME
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



