Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Methanol ≥99.6%, Laboratory Reagent

Glass bottle, 4 x 2.5 l, 10 L
Codice catalogo 179957-4X2.5L
Health
Flammable
Nome del fornitore:   Honeywell Chemicals
Methanol ≥99.6%, Laboratory Reagent
Codice catalogo 179957-4X2.5L
Nome del fornitore:  Honeywell Chemicals
Numero CAS:  67-56-1
Methanol ≥99.6%, Laboratory Reagent
Methanol ≥99.6%, Laboratory Reagent

Glass bottle, 4 x 2.5 l, 10 L

Numero CAS:  67-56-1
Price
144,25 €
Log in for availability
1 * 10 L
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. Ulteriori informazioni
Product Details & Documents
  • ≥95% for general laboratory use.

Beilstein Number: 1098229

Synonyms:  Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
  • CAS Number:67-56-1
  • Formula:CH₃OH
  • MDL Number:MFCD00004595
  • Molecular Weight:32.04 g/mol
  • Merck Index:13,05984
  • Boiling Pt:64…65 °C (1.013 hPa)
  • Melting Pt:–98 °C
  • Density:0.79 g/cm³ (20 °C)
  • Flash Pt:11 °C
  • Storage Temperature:Ambient
  • EINECS:200-659-6
  • UN:1230
  • ADR:3,II
  • REACH:01-2119433307-44
Specifications

Specification Test Results

  • Explosion Limit:
    36 %
  • Identity (IR):
    complying
  • Assay (GC):
    Min. 99.6 %
  • Non-volatile matter:
    Max. 0.01 %
  • Water (Karl Fischer):
    Max. 0.1 %
  • Free acid (as HCOOH):
    Max. 0.03 %
  • Free alkali (as NH₃):
    Max. 0.01 %
  • Vapor Pressure:
    410 mmHg ( 50 °C)
  • Autoignition Temperature:
    725 °F
  • Vapor Density:
    1.11 (vs air)

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 L
  • Pk Info:
    4 x 2.5 l

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.