Inserisci ordine
Italy
Orders LinkContactUsLinkComponent
 

Titanium Standard for ICP, TraceCERT®, Supelco®

Titanio, Ti, 1000 mg/L, Nitric acid dil., 100 mL
Catalog # 12237-100ML
Supplier:   Merck
Titanium Standard for ICP, TraceCERT®, Supelco®
Catalog # 12237-100ML
Supplier:  Merck
CAS Number:  
Titanium Standard for ICP, TraceCERT®, Supelco®
Titanium Standard for ICP, TraceCERT®, Supelco®

Titanio, Ti, 1000 mg/L, Nitric acid dil., 100 mL

CAS Number:  
Price
123,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

 : This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034

 

Specifications

Specifications

  • Concentrazione:
    1000 mg/L
  • Matrice:
    Nitric acid dil.
  • Conf:
    100 mL
  • Componenti:
    Titanio
  • Simbolo:
    Ti
  • Cat. No.:
    12237-100ML
  • M-Clarity quality level:
    MQ300

Specifications