Order Entry
Italy
Orders LinkContactUsLinkComponent
Allyl alcohol ≥98%

Allyl alcohol ≥98%

Glass bottle, 1.000 mL
Catalog # A15026.AU
Flammable
Toxic
Irritant
Allyl alcohol ≥98%
Catalog # A15026.AU
CAS Number:  107-18-6
Allyl alcohol ≥98%
Allyl alcohol ≥98%

Glass bottle, 1.000 mL

CAS Number:  107-18-6
Price
76,70 €
Log in for availability
1 * 1.000 mL
Product Details & Documents

Purity: 98+% CAS No.: 107-18-6 Molecular Formula: C3H6O Formula Weight: 58.08 Flash Point: 22°(71°F) Refractive Index: 1.4130 Odour: Pungent Melting Point: -129° Boiling Point: 95-97° Beilstein Reference: 605307 Merck Reference: 14,286 UN No.: 1098 MDL No.: MFCD00002920

Synonyms:  3-Hydroxypropene, 2-Propen-1-ol
  • CAS Number:107-18-6
  • Formula:C₃H₆O
  • MDL Number:MFCD00002920
  • Molecular Weight:58.08 g/mol
  • Merck Index:14,286
  • Boiling Pt:95…97 °C
  • Melting Pt:–129 °C
  • Density:0.852
  • Flash Pt:22 °C (71 °F)
  • Storage Temperature:Ambient
  • EINECS:203-470-7
  • UN:1098
  • ADR:6.1,I
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1.000 mL

Specifications

Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..