Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Ribbons for label printer BBP™11

Thermal transfer printer ribbon, width 110 mm, BBP™11, 70 m, Black
Codice catalogo BRDY804465
Nome del fornitore:   Brady
Ribbons for label printer BBP™11
Codice catalogo BRDY804465
Nome del fornitore:  Brady
Ribbons for label printer BBP™11
Ribbons for label printer BBP™11

Thermal transfer printer ribbon, width 110 mm, BBP™11, 70 m, Black

Price
48,20 €
Log in for availability
1 * 1 Roll
Product Details & Documents
Specifications
  • Colour:
    Black
  • Description:
    Thermal transfer printer ribbon, width 110 mm
  • Length:
    70 m
  • Seq:
    18
  • For:
    BBP™11