Accessories for wire baskets
575×375 mm, mesh dimensions: 35×35×3 mm
Catalog # BOCH17325
Supplier: BOCHEM
Jump to:
Product Details & Documents
Specifications
- Description:575×375 mm, mesh dimensions: 35×35×3 mm
- Seq:15
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



