Order Entry
Italy
Orders LinkContactUsLinkComponent
Accessories for wire baskets

Accessories for wire baskets

275×375 mm, mesh dimensions: 10×10×3 mm
Catalog # BOCH17316
Supplier:   BOCHEM
Accessories for wire baskets
Catalog # BOCH17316
Supplier:  BOCHEM
Accessories for wire baskets
Accessories for wire baskets

275×375 mm, mesh dimensions: 10×10×3 mm

Price
414,00 €
Log in for availability
1 * 1 pezzi
Product Details & Documents
Specifications
  • Description:
    275×375 mm, mesh dimensions: 10×10×3 mm
  • Seq:
    10
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.