Support clamps
DN 60, Schott laboratory flat flanges (PF)
Catalog # BOCH17210
Supplier: BOCHEM
Jump to:
Product Details & Documents
Support clamps made of stainless steel (18/19) and PTFE for either Schott laboratory flat flanges (PF) (optimal flange angle 45°) or Schott "Kugel-Flanges" (ball and socket system) (KF).
- For reactors and other vessels
- User friendly thanks to joint screws
- With two supporting devices for 26,9 mm tubes
Specifications
- For:Schott laboratory flat flanges (PF)
- Description:DN 60
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



