Order Entry
Italy
Orders LinkContactUsLinkComponent
Scalpels
Scalpels
3, 150 mm, Scalpel, with mounted blade, orange
Codice catalogo BOCH12642
Nome del fornitore:   BOCHEM
Scalpels
Codice catalogo BOCH12642
Nome del fornitore:  BOCHEM
Scalpels
Scalpels
3, 150 mm, Scalpel, with mounted blade, orange
Price
4,20 €
Log in for availability
1 * 1 pezzi
Product Details & Documents
These mounted scalpels feature a steel blade and a coloured handle made of impact resistant PS.
  • Not sterile
  • Blades are exchangeable
  • Spare blades available (set of 10 pieces)
Specifications
  • Type:
    3
  • Description:
    Scalpel, with mounted blade, orange
  • Length:
    150 mm