Water purification systems, Select Analyst
Select Analyst 320 GP (without internal tank)
171-0281
The Select Analyst is a compact unit ideal for Laboratory use, delivering a steady supply of > 1MΩ.cm pure water for Laboratory tasks using carbon pre-treatment, reverse osmosis and deionisation.
- RO removes > 98% minerals and > 99% bacteria
- RO boost pump fitted as standard
- LCD colour touch screen panel, visual and audible alarms
- Available with or without integral 20 litre storage
- Semi-automatic clean cycle
- USB port to download event data and upload software updates
- WiFi connection
Typical applications include glassware rinsing, buffers and stains, reagent make-up, and media preparation.
Installation kit and all consumables included for first year's operation.
: Supplied with alphanumeric display panel, pretreatment cartridge, reverse osmosis module, deionisation module and single tank bib-tap. External tanks required for GP models are also available, please contact your local Avantor office for details.
- Description:Select Analyst 320 GP (without internal tank)
- Seq:11
- Cat. No.:171-0281
- Production flow rate:30 - 48 L/h
- Feed water:Potable (tap)
- Model:Analyst 320
- Bacteria:>99% rejection*
- W×D×H:440×560×750 mm
- TOC content:<50 ppb
- Weight:33 - 52 kg
- Inorganics @ 25°C:>1 MΩ/cm
Accessories
- Description
- Description
- 171-0614External tank wall mounting kit, for Select Analyst and HP systems1 * 1 pezzi - 1 410,00 €
- Description
- Description
- 171-0256Annual consumable pack for Select Analyst 320/Purewater 300 containing 2 x PT16 and 2 x PP16 cartridges1 * 1 pezzi - 4 120,00 €
- Description
- Description
- 171-0265Filter element, 0,2 µm for current Select range (with grey door) and previous L2G Select range (with blue door), used in HP, Fusion, Neptune water purification systems1 * 1 pezzi - 769,00 €
- Description
- Description
- Description
- Description



