Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Water purification systems, Select Analyst

Select Analyst 160 GP with boost pump, without internal tank (external tank must be ordered separately)
  171-0280
 :   Purite
Water purification systems, Select Analyst
  171-0280
 :  Purite
Water purification systems, Select Analyst
Water purification systems, Select Analyst

Select Analyst 160 GP with boost pump, without internal tank (external tank must be ordered separately)

 
10 970,00 €
   
1 * 1 pezzi
 

The Select Analyst is a compact unit ideal for Laboratory use, delivering a steady supply of > 1MΩ.cm pure water for Laboratory tasks using carbon pre-treatment, reverse osmosis and deionisation.

  • RO removes > 98% minerals and > 99% bacteria
  • RO boost pump fitted as standard
  • LCD colour touch screen panel, visual and audible alarms
  • Available with or without integral 20 litre storage
  • Semi-automatic clean cycle
  • USB port to download event data and upload software updates
  • WiFi connection

Typical applications include glassware rinsing, buffers and stains, reagent make-up, and media preparation.

Installation kit and all consumables included for first year's operation.

 : Supplied with alphanumeric display panel, pretreatment cartridge, reverse osmosis module, deionisation module and single tank bib-tap. External tanks required for GP models are also available, please contact your local Avantor office for details.

 
  • Description:
    Select Analyst 160 GP with boost pump, without internal tank (external tank must be ordered separately)
  • Seq:
    09
  • Cat. No.:
    171-0280
  • Production flow rate:
    14,4 - 24 L/h
  • Feed water:
    Potable (tap)
  • Model:
    Analyst 160
  • Bacteria:
    >99% rejection*
  • W×D×H:
    440×560×750 mm
  • TOC content:
    <50 ppb
  • Weight:
    28 - 43 kg
  • Inorganics @ 25°C:
    >1 MΩ/cm
Accessories

 

  •  
    Description
     
     
  • 171-0613
    External tank 50 l, for Select Analyst systems
       
    1 * 1 pezzi - 2 420,00 €

 

  •  
    Description
     
     
  • 171-0614
    External tank wall mounting kit, for Select Analyst and HP systems
       
    1 * 1 pezzi - 1 410,00 €

 

  •  
    Description
     
     
  • 171-0612
    Wall mounting bracket for Select systems
       
    1 * 1 pezzi

 

  •  
    Description
     
     
  • 171-0256
    Annual consumable pack for Select Analyst 320/Purewater 300 containing 2 x PT16 and 2 x PP16 cartridges
       
    1 * 1 pezzi - 4 120,00 €

 

  •  
    Description
     
     
  • 171-0878
    Remote display for Select Analyst, PW300 systems
       
    1 * 1 pezzi - 580,00 €

 

  •  
    Description
     
     
  • 171-0265
    Filter element, 0,2 µm for current Select range (with grey door) and previous L2G Select range (with blue door), used in HP, Fusion, Neptune water purification systems
       
    1 * 1 pezzi - 769,00 €

 

  •  
    Description
     
     
  • 171-0264
    Replacement dispense gun for water purification systems
       
    1 * 1 pezzi - 732,00 €

 

  •  
    Description
     
     
  • 171-0883
    External tank 100 l, for Select Analyst systems
       
    1 * 1 pezzi - 3 120,00 €

 

  •  
    Description
     
     
  • 171-0884
    Wall mounting kit for Select systems
       
    1 * 1 pezzi - 1 330,00 €

 

  •  
    Description
     
     
  • 171-0877
    Remote dispense for Select Analyst, HP, Fusion and Neptune systems
       
    1 * 1 pezzi - 869,00 €