Order Entry
Italy
Orders LinkContactUsLinkComponent
Microplates, 96-well
Microplates, 96-well
96 Well plate, Translucent, 96, -
Codice catalogo RATI6018121
Nome del fornitore:   RATIOLAB
Microplates, 96-well
Codice catalogo RATI6018121
Nome del fornitore:  RATIOLAB
Microplates, 96-well
Microplates, 96-well
96 Well plate, Translucent, 96, -
Price
63,60 €
Log in for availability
1 * 100 pezzi
Product Details & Documents
Specifications
  • No. of wells:
    96
  • Sterile:
    -
  • Colour:
    Translucent
  • Description:
    96 Well plate
  • Cat. No.:
    RATI6018121
  • Model:
    Micro Test Plates, volume ∼300 μl
  • Well profile:
    U-bottom