Order Entry
Italy
Orders LinkContactUsLinkComponent
Microplates, 96-well
Microplates, 96-well
96 Well plate, Clear, 96, -
Catalog # RATI6018211
Supplier:   RATIOLAB
Microplates, 96-well
Catalog # RATI6018211
Supplier:  RATIOLAB
Microplates, 96-well
Microplates, 96-well
96 Well plate, Clear, 96, -
Price
63,60 €
Log in for availability
1 * 100 pezzi
Product Details & Documents
Specifications
  • No. of wells:
    96
  • Sterile:
    -
  • Colour:
    Clear
  • Description:
    96 Well plate
  • Cat. No.:
    RATI6018211
  • Model:
    Micro Test Plates, volume ∼300 μl
  • Well profile:
    F-bottom