Phosphate buffer solution, pH 7,2 ( 25 °C), Fluka™
Phosphate buffer solution, 7,20, ±0,04, Plastic Bottle, 2 L
Catalog # 319252-2L
Supplier: Honeywell Chemicals
Phosphate buffer solution, pH 7,2 ( 25 °C), Fluka™
Catalog # 319252-2L
Supplier: Honeywell Chemicals
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Plastic Bottle
- Pk:2 L
- Description:Phosphate buffer solution
- pH accuracy:±0,04
- pH value:7,20



