Order Entry
Italy
Orders LinkContactUsLinkComponent
dNTP solutions, illustra™
dCTP, 100 mMol solution, 100 µmol
  28-4065-12
 :   Cytiva
dNTP solutions, illustra™
  28-4065-12
 :  Cytiva
 :  
dNTP solutions, illustra™
dNTP solutions, illustra™
dCTP, 100 mMol solution, 100 µmol
 :  
170,00 €
1 * 100 umol
illustra™ dNTPs are high purity solutions for enzymatically sensitive applications, such as nucleic acid amplification, sequencing, labelling, mutagenesis, cDNA synthesis and expression profiling.
  • Free from DNase, RNase, and nicking enzyme activity
  • Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
  • Buffer free and ready-to-use solutions in multiple formats
  • Functionally tested for long PCR and sequencing
  • Pk:
    100 µmol
  • Description:
    dCTP, 100 mMol solution