dNTP solutions, illustra™
dUTP, 100 mM solution, 25 µmol
Catalog # 28-4065-41
Supplier: Cytiva
Jump to:
Product Details & Documents
illustra™ dNTPs are high purity solutions for enzymatically sensitive applications, such as nucleic acid amplification, sequencing, labelling, mutagenesis, cDNA synthesis and expression profiling.
- Free from DNase, RNase, and nicking enzyme activity
- Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
- Buffer free and ready-to-use solutions in multiple formats
- Functionally tested for long PCR and sequencing
Specifications
Specifications
- Pk:25 µmol
- Description:dUTP, 100 mM solution