Order Entry
Italy
ContactUsLinkComponent
Rack, flipper 1, colour orange, material PP, autoclavable, temperature resistant -90°C to +120°C, dimension 197x95x115mm, 8 holes di ameter 30mm, 20 holes diameter 20mm. 1 * 1 items
Catalog # KOEH2-2774
Nome del fornitore:  KOEHLER TECHNISCHE PRODUKTEN
Rack, flipper 1, colour orange, material PP, autoclavable, temperature resistant -90°C to +120°C, dimension 197x95x115mm, 8 holes di ameter 30mm, 20 holes diameter 20mm. 1 * 1 items
Catalog # KOEH2-2774
Nome del fornitore:  KOEHLER TECHNISCHE PRODUKTEN
Frequently Bought Together