Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Capillary columns, Chromolith® CapRod®

Capillary, RP-18e HR, 0,20 mm, 150 mm
Catalog # 1.50407.0001
Supplier:   Merck
Capillary columns, Chromolith® CapRod®
Catalog # 1.50407.0001
Supplier:  Merck
Capillary columns, Chromolith® CapRod®
Capillary columns, Chromolith® CapRod®

Capillary, RP-18e HR, 0,20 mm, 150 mm

Price
Inquire For Price
Log in for availability
1 * 1 pezzi
Product Details & Documents

Fast nano-LC for proteins and peptides. Chromolith® CapRod® capillaries combine the speed of monolithic silica technology with the sensitivity of nano-LC, thus enabling high speed high sensitivity LC-MS analytics.

  • High flow rates at low pressure
  • Long column lifetime
  • Robust handling
Specifications
  • Column Format:
    Capillary
  • Ø int.:
    0,20 mm
  • Length:
    150 mm
  • Seq:
    08
  • Phase:
    RP-18e HR