Housing lower part (Polymax 2040 / Promax 2020 / Unimax 2010) 1 * 1 items
Catalog # HEID23-07-08-01-49
Supplier: Heidolph Instruments GmbH & Co.KG
Housing lower part (Polymax 2040 / Promax 2020 / Unimax 2010) 1 * 1 items
Catalog # HEID23-07-08-01-49
Supplier: Heidolph Instruments GmbH & Co.KG
Frequently Bought Together
Accessories for Titrette®
Supplier: Brand
Verdichter NEK 6160Z-267BA (50 Hz) 1 * 1 items
Supplier: Binder
LEVEL INDICATOR 1 * 1 items
Supplier: Sartorius Balances
Accessories for multi channel pipettes, mechanical, Research® plus
Supplier: EPPENDORF
BIOTECTOR ACID REAGENT WITH CATALYST 1 * 20 L
Supplier: Hach
Tip cone mLINE 1000µl (chemically resistant) 1 * 1 items
Supplier: Sartorius
Vacuum seal PTFE / FKM
Supplier: Heidolph Instruments GmbH & Co.KG
Handheld refractometers, Eclipse
Supplier: BELLINGHAM STANLEY
Casing (Polymax 2040 / Promax 2020 / Unimax 2010) 1 * 1 EU
Supplier: Heidolph Instruments GmbH & Co.KG
Accessories for water purification millipore
Supplier: Merck
O-Ring 120x3 for filter exhaust air filter housing 1 * 1 items
Supplier: SHP Steriltechnik
V-ring for x200lvr & x300lvr (x24) 1 * 1 items
Supplier: GILSON
Accessories for Lauda ECO range circulators
Supplier: LAUDA
Accessories for autoclaves EL/ELV and D-Line
Supplier: TUTTNAUER