Order Entry
Italy
Orders LinkContactUsLinkComponent
Pipette tips, Finntip™ Flex

Pipette tips, Finntip™ Flex

1 - 200 µl, -, 60 mm, Yellow, 10 racks, 96 each
Catalog # 613-0797
   Low Manufacturing Impact
Pipette tips, Finntip™ Flex
Catalog # 613-0797
   Low Manufacturing Impact
Pipette tips, Finntip™ Flex
Pipette tips, Finntip™ Flex

1 - 200 µl, -, 60 mm, Yellow, 10 racks, 96 each

Price
73,00 €
Sale

Valid until 30/06/2026

Log in for availability
1 * 960 pezzi
Product Details & Documents

New generation of tips offering the user greater flexibility.

  • High quality PP, autoclavable
  • Ergonomic thin wall tip; low attachment and ejection forces
  • Non slip rack with hinged cover
  • Easy to use refill system

Packaging: Available loose, in racks, in a starter kit (1 rack of 96 and 16 or 20 refills of 96) or a refill kit (16 or 20 refills of 96).
10 ml tips: only available in bags.

Specifications
  • Colour code:
    Yellow
  • Packed:
    10 racks, 96 each
  • Sterile:
    -
  • Volume:
    1 - 200 µl
  • Length:
    60 mm
  • Cat. No.:
    613-0797
  • Environmental certification:
    ISO 14001 (Certificate Registration No. EMS 623400)
  • Environmentally Preferable:
    Low Manufacturing Impact
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.