Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Water purification systems, Select Analyst

Select Analyst 320 with internal tank (30 L)
Catalog # 171-0188
Water purification systems, Select Analyst
Catalog # 171-0188
Supplier:  
Water purification systems, Select Analyst
Water purification systems, Select Analyst

Select Analyst 320 with internal tank (30 L)

Discontinued
Product Details & Documents

The Select Analyst is a compact unit ideal for Laboratory use, delivering a steady supply of > 1MΩ.cm pure water for Laboratory tasks using carbon pre-treatment, reverse osmosis and deionisation.

  • RO removes > 98% minerals and > 99% bacteria
  • RO boost pump fitted as standard
  • LCD colour touch screen panel, visual and audible alarms
  • Available with or without integral 20 litre storage
  • Semi-automatic clean cycle
  • USB port to download event data and upload software updates
  • WiFi connection

Typical applications include glassware rinsing, buffers and stains, reagent make-up, and media preparation.

Installation kit and all consumables included for first year's operation.

 : Supplied with alphanumeric display panel, pretreatment cartridge, reverse osmosis module, deionisation module and single tank bib-tap. External tanks required for GP models are also available, please contact your local Avantor office for details.

Specifications
  • Description:
    Select Analyst 320 with internal tank (30 L)
  • Seq:
    12
  • Cat. No.:
    171-0188
  • Production flow rate:
    30 - 48 L/h
  • Feed water:
    Potable (tap)
  • Model:
    Analyst 320
  • Bacteria:
    >99% rejection*
  • W×D×H:
    440×560×750 mm
  • TOC content:
    <50 ppb
  • Weight:
    33 - 52 kg
  • Inorganics @ 25°C:
    >1 MΩ/cm
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • External tank 50 l, for Select Analyst systems
    External tank 50 l, for Select Analyst systems
    171-0613
    Log in for availability
    1 * 1 pezzi - 2 420,00 €

Product Information

  • Catalog #
    Availability
    Price
  • External tank wall mounting kit, for Select Analyst and HP systems
    External tank wall mounting kit, for Select Analyst and HP systems
    171-0614
    Log in for availability
    1 * 1 pezzi - 1 410,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Wall mounting bracket for Select systems
    Wall mounting bracket for Select systems
    171-0612
    Log in for availability
    1 * 1 pezzi

Product Information

  • Catalog #
    Availability
    Price
  • Annual consumable pack for Select Analyst 320/Purewater 300 containing 2 x PT16 and 2 x PP16 cartridges
    Annual consumable pack for Select Analyst 320/Purewater 300 containing 2 x PT16 and 2 x PP16 cartridges
    171-0256
    Log in for availability
    1 * 1 pezzi - 4 120,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Remote display for Select Analyst, PW300 systems
    Remote display for Select Analyst, PW300 systems
    171-0878
    Log in for availability
    1 * 1 pezzi - 580,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Filter element, 0,2 µm for current Select range (with grey door) and previous L2G Select range (with blue door), used in HP, Fusion, Neptune water purification systems
    Filter element, 0,2 µm for current Select range (with grey door) and previous L2G Select range (with blue door), used in HP, Fusion, Neptune water purification systems
    171-0265
    Log in for availability
    1 * 1 pezzi - 769,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Replacement dispense gun for water purification systems
    Replacement dispense gun for water purification systems
    171-0264
    Log in for availability
    1 * 1 pezzi - 732,00 €

Product Information

  • Catalog #
    Availability
    Price
  • External tank 100 l, for Select Analyst systems
    External tank 100 l, for Select Analyst systems
    171-0883
    Log in for availability
    1 * 1 pezzi - 3 120,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Wall mounting kit for Select systems
    Wall mounting kit for Select systems
    171-0884
    Log in for availability
    1 * 1 pezzi - 1 330,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Remote dispense for Select Analyst, HP, Fusion and Neptune systems
    Remote dispense for Select Analyst, HP, Fusion and Neptune systems
    171-0877
    Log in for availability
    1 * 1 pezzi - 869,00 €