Water purification systems, Select Analyst
Product Details & Documents
The Select Analyst is a compact unit ideal for Laboratory use, delivering a steady supply of > 1MΩ.cm pure water for Laboratory tasks using carbon pre-treatment, reverse osmosis and deionisation.
- RO removes > 98% minerals and > 99% bacteria
- RO boost pump fitted as standard
- LCD colour touch screen panel, visual and audible alarms
- Available with or without integral 20 litre storage
- Semi-automatic clean cycle
- USB port to download event data and upload software updates
- WiFi connection
Typical applications include glassware rinsing, buffers and stains, reagent make-up, and media preparation.
Installation kit and all consumables included for first year's operation.
Delivery information: Supplied with alphanumeric display panel, pretreatment cartridge, reverse osmosis module, deionisation module and single tank bib-tap. External tanks required for GP models are also available, please contact your local Avantor office for details.
Specifications
- Description:Select Analyst 80 IT with boost pump and internal tank (20 L)
- Seq:05
- Cat. No.:171-0185
- Production flow rate:7,2 - 12 L/h
- Feed water:Potable (tap)
- Model:Analyst 80
- Bacteria:>99% rejection*
- W×D×H:440×560×750 mm
- TOC content:<50 ppb
- Weight:28 - 43 kg
- Inorganics @ 25°C:>1 MΩ/cm
Accessories
Product Information
- Catalog #AvailabilityPrice
- External tank 50 l, for Select Analyst systemsExternal tank 50 l, for Select Analyst systems171-0613Log in for availability1 * 1 pezzi - 2 420,00 €
Product Information
- Catalog #AvailabilityPrice
- External tank wall mounting kit, for Select Analyst and HP systemsExternal tank wall mounting kit, for Select Analyst and HP systems171-0614Log in for availability1 * 1 pezzi - 1 410,00 €
Product Information
- Catalog #AvailabilityPrice
- Wall mounting bracket for Select systems
Product Information
- Catalog #AvailabilityPrice
- Annual consumable pack for Select Analyst 320/Purewater 300 containing 2 x PT16 and 2 x PP16 cartridgesAnnual consumable pack for Select Analyst 320/Purewater 300 containing 2 x PT16 and 2 x PP16 cartridges171-0256Log in for availability1 * 1 pezzi - 4 120,00 €
Product Information
- Catalog #AvailabilityPrice
- Remote display for Select Analyst, PW300 systemsRemote display for Select Analyst, PW300 systems171-0878Log in for availability1 * 1 pezzi - 580,00 €
Product Information
- Catalog #AvailabilityPrice
- Filter element, 0,2 µm for current Select range (with grey door) and previous L2G Select range (with blue door), used in HP, Fusion, Neptune water purification systemsFilter element, 0,2 µm for current Select range (with grey door) and previous L2G Select range (with blue door), used in HP, Fusion, Neptune water purification systems171-0265Log in for availability1 * 1 pezzi - 769,00 €
Product Information
- Catalog #AvailabilityPrice
- Replacement dispense gun for water purification systemsReplacement dispense gun for water purification systems171-0264Log in for availability1 * 1 pezzi - 732,00 €
Product Information
- Catalog #AvailabilityPrice
- External tank 100 l, for Select Analyst systemsExternal tank 100 l, for Select Analyst systems171-0883Log in for availability1 * 1 pezzi - 3 120,00 €
Product Information
- Catalog #AvailabilityPrice
- Wall mounting kit for Select systems
Product Information
- Catalog #AvailabilityPrice
- Remote dispense for Select Analyst, HP, Fusion and Neptune systemsRemote dispense for Select Analyst, HP, Fusion and Neptune systems171-0877Log in for availability1 * 1 pezzi - 869,00 €
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



