Stopcocks, Rotaflo®
3 mm, 10 mm
Catalog # BIBBHP3RA/7
Supplier: SciLabware
Jump to:
Product Details & Documents
PYREX®, borosilicate glass with PTFE needle valve.
- Lubricant is not required (no seizure, no contamination with grease)
- Interchangeable elements
- Precise flow control
These chemically inert stopcocks can be used at temperatures up to +200 °C. Two different models are available: general purpose (vacuum rating up to 10-4 mm Hg) and high performance (vacuum rating up to 10-9 mm Hg).
Specifications
- Hole Ø:3 mm
- Seq:15
- For tube I-Ø:10 mm
Accessories
Product Information
- Catalog #ForAvailabilityPrice
- Replacement key, bore Ø 6 mmReplacement key, bore Ø 6 mm229-0230High performance Rotaflo® stopcocksLog in for availability1 * 2 pezzi - 141,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Replacement key, bore Ø 6 mmReplacement key, bore Ø 6 mm229-0231General purpose Rotaflo® stopcocksLog in for availability1 * 5 pezzi - 194,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Replacement locking ring for key with bore 6 mmReplacement locking ring for key with bore 6 mmBIBBLR6Rotaflo® stopcocksLog in for availability1 * 5 pezzi - 46,50 €
Product Information
- Catalog #ForAvailabilityPrice
- Replacement key, bore Ø 10 mmReplacement key, bore Ø 10 mm229-0226High performance Rotaflo® stopcocksLog in for availability1 * 1 pezzi - 90,40 €
Product Information
- Catalog #ForAvailabilityPrice
- Replacement key, bore Ø 3 mmReplacement key, bore Ø 3 mm229-0229High performance Rotaflo® stopcocksLog in for availability1 * 2 pezzi - 101,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Replacement locking ring for key with bore 3 mmReplacement locking ring for key with bore 3 mmBIBBLR3Rotaflo® stopcocksLog in for availability1 * 5 pezzi - 31,70 €
Product Information
- Catalog #ForAvailabilityPrice
- Replacement key, bore Ø 10 mmReplacement key, bore Ø 10 mm229-0234General purpose Rotaflo® stopcocksLog in for availability1 * 5 pezzi - 303,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Replacement key, bore Ø 3 mmReplacement key, bore Ø 3 mmBIBBGP3KGeneral purpose Rotaflo® stopcocksLog in for availability1 * 5 pezzi - 125,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Replacement locking ring for key with bore 10 mmReplacement locking ring for key with bore 10 mmBIBBLR10Rotaflo® stopcocksLog in for availability1 * 5 pezzi - 43,40 €