Order Entry
Italy
Orders LinkContactUsLinkComponent
Spore strips

Spore strips

Geobacillus stearothermophilus cell line 7953: mini strip 106 (24 month shelf life) / sterilisation process: steam, 10 mm, 2 mm
Catalog # NAMSSTS-062B
Supplier:   CROSSTEX
Spore strips
Catalog # NAMSSTS-062B
Supplier:  CROSSTEX
Spore strips

Geobacillus stearothermophilus cell line 7953: mini strip 106 (24 month shelf life) / sterilisation process: steam, 10 mm, 2 mm

Price
675,00 €
Log in for availability
1 * 100 pezzi
Product Details & Documents

Documents

Specifications
  • Width:
    2 mm
  • Description:
    Geobacillus stearothermophilus cell line 7953: mini strip 106 (24 month shelf life) / sterilisation process: steam
  • Length:
    10 mm
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.