Order Entry
Italy
Orders LinkContactUsLinkComponent
Tris(3-chlorophenyl)phosphine ≥98%

Tris(3-chlorophenyl)phosphine ≥98%

Glass bottle for solids, 5 g
Codice catalogo A11203.06
Irritant
Nome del fornitore:   Thermo Fisher Scientific
Tris(3-chlorophenyl)phosphine ≥98%
Codice catalogo A11203.06
Nome del fornitore:  Thermo Fisher Scientific
Numero CAS:  29949-85-7
Tris(3-chlorophenyl)phosphine ≥98%
Tris(3-chlorophenyl)phosphine ≥98%

Glass bottle for solids, 5 g

Numero CAS:  29949-85-7
Price
132,00 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 98% CAS No.: 29949-85-7 Molecular Formula: C18H12Cl3P Formula Weight: 365.63 Melting Point: 64-66° MDL No.: MFCD00013632

Synonyms:  Tri(m-chlorophenyl)phosphine, Tris(m-chlorophenyl)phosphine
  • CAS Number:29949-85-7
  • Formula:C₁₈H₁₂Cl₃P
  • MDL Number:MFCD00013632
  • Molecular Weight:365.63 g/mol
  • Melting Pt:64…66 °C
  • Storage Temperature:Ambient
  • EINECS:249-970-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications