Tris(3-chlorophenyl)phosphine ≥98%
Glass bottle for solids, 1 g
Purity: 98% CAS No.: 29949-85-7 Molecular Formula: C18H12Cl3P Formula Weight: 365.63 Melting Point: 64-66° MDL No.: MFCD00013632
: Tri(m-chlorophenyl)phosphine, Tris(m-chlorophenyl)phosphine
- :29949-85-7
- :C₁₈H₁₂Cl₃P
- :MFCD00013632
- :365.63 g/mol
- :64…66 °C
- :Ambient
- :249-970-9
- Pack type:Glass bottle for solids
- Pk:1 g



