Order Entry
Italy
Orders LinkContactUsLinkComponent
2-Chlorothioxanthone ≥99%

2-Chlorothioxanthone ≥99%

Glass bottle for solids, 25 g
Catalog # A18131.14
2-Chlorothioxanthone ≥99%
Catalog # A18131.14
CAS Number:  86-39-5
2-Chlorothioxanthone ≥99%
2-Chlorothioxanthone ≥99%

Glass bottle for solids, 25 g

CAS Number:  86-39-5
Price
129,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 99% CAS No.: 86-39-5 Molecular Formula: C13H7ClOS Formula Weight: 246.72 Flash Point: 91°(196°F) Melting Point: 151-154° MDL No.: MFCD00005067

Synonyms:  2-Chlorothioxanthen-9-one, 2-Chloro-9H-thioxanthen-9-one
  • CAS Number:86-39-5
  • Formula:C₁₃H₇ClOS
  • MDL Number:MFCD00005067
  • Molecular Weight:246.72 g/mol
  • Melting Pt:151…154 °C
  • Flash Pt:91 °C (196 °F)
  • Storage Temperature:Ambient
  • EINECS:201-667-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications