Order Entry
Italy
Orders LinkContactUsLinkComponent
Stands for MSC-Advantage
Manually adjustable floor stand, 750 to 950 mm working height in 50 mm increments, for 1,8 m cabinet
Catalog # 135-1317
Supplier:   Thermo Scientific
Stands for MSC-Advantage
Catalog # 135-1317
Supplier:  Thermo Scientific
Stands for MSC-Advantage
Stands for MSC-Advantage
Manually adjustable floor stand, 750 to 950 mm working height in 50 mm increments, for 1,8 m cabinet
Price
1 440,00 €
Log in for availability
1 * 1 pezzi
Product Details & Documents
Specifications
  • Description:
    Manually adjustable floor stand, 750 to 950 mm working height in 50 mm increments, for 1,8 m cabinet
  • Seq:
    08