Order Entry
Italy
Orders LinkContactUsLinkComponent
Bottles, water sampling
Bottles, water sampling
500 ml, single wrapped
Catalog # 300-0161
Supplier:   LP ITALIANA
Bottles, water sampling
Catalog # 300-0161
Supplier:  LP ITALIANA
Bottles, water sampling
Bottles, water sampling
500 ml, single wrapped
Price
540,00 €
Log in for availability
1 * 120 pezzi
Product Details & Documents
PP, translucent, square, with screw cap.
  • Tamper evident screw caps
  • Sterile
  • Moulded graduations
Bottles for microbiological analysis of chlorinated or non-chlorinated water. Sodium thiosulphate inhibits the effects of chlorine, bromide and ozone present in the water. Ideal for flat storage (fridges, etc).
 : *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
  • Packed:
    single wrapped
  • Capacity:
    500 ml
  • Seq:
    04